1-Bromo-3-methylimidazo[1,5-a]pyrazine 98%

Supplier: Ambeed
Warning

A215587-250MG A215587-100MG A215587-5G
76814-732EA 55 USD
76814-732 77883-532 77883-533
1-Bromo-3-methylimidazo[1,5-a]pyrazine 98%
1-Bromo-3-methylimidazo[1,5-a]pyrazine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MDL Number: MFCD09999201
CAS Number: 56481-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR