1,3-Dimethyl-1H-benzimidazol-3-ium iodide 97%

Supplier: Ambeed
Warning

A215560-100MG A215560-1G A215560-250MG A215560-5G
76814-726EA 12 USD
76814-726 76814-728 76814-730 77883-531
1,3-Dimethyl-1H-benzimidazol-3-ium iodide 97%
1,3-Dimethyl-1H-benzimidazol-3-ium iodide

Precaución: for Research Use Only

Formula: C₉H₁₁N₂·I
MDL Number: MFCD00491277
CAS Number: 7181-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR