2,6-Difluoro-3-formylphenylboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: (2,6-Difluoro-3-formylphenyl)boronic acid

A215250-1G A215250-250MG A215250-5G
76814-702EA 81.69 USD
76814-702 76814-704 76814-706
2,6-Difluoro-3-formylphenylboronic acid 98%
2,6-Difluoro-3-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₃
MW: 185.9200 g/mol
MDL Number: MFCD07369756
CAS Number: 849062-09-5
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR