6-Chloro-2-nitropyridin-3-ol 98%

Supplier: Ambeed

A215108-100MG A215108-1G A215108-250MG A215108-5G
76814-686EA 25.47 USD
76814-686 76814-688 76814-690 76814-692
6-Chloro-2-nitropyridin-3-ol 98%
6-Chloro-2-nitropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₃
MW: 174.54 g/mol
MDL Number: MFCD09834122
CAS Number: 887471-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR