3,5-Dibromoisonicotinaldehyde 98%

Supplier: Ambeed
Warning

A214792-10G A214792-1G A214792-5G A214792-25G A214792-100G
76814-680EA 14 USD
76814-680 76814-682 76814-684 77612-112 77671-914
3,5-Dibromoisonicotinaldehyde 98%
3,5-Dibromoisonicotinaldehyde

Precaución: for Research Use Only

Formula: C6H3Br2NO
MDL Number: MFCD06410681
CAS Number: 70201-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR