Thiophen-3-amine hydrochloride 95%

Supplier: Ambeed
Danger

A214720-5G A214720-10G A214720-1G A214720-25G
76814-678EA 34 USD
76814-678 77568-072 77568-074 77671-912
Thiophen-3-amine hydrochloride 95%
Thiophen-3-amine hydrochloride

Precaución: For research use only.

Formula: C₄H₆ClNS
MDL Number: MFCD11226843
CAS Number: 25475-76-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR