4-Methyl-5-nitrobenzene-1,2-diamine 95%

Supplier: Ambeed
Warning

A214464-100MG A214464-250MG
76814-672EA 63.66 USD
76814-672 76814-674
4-Methyl-5-nitrobenzene-1,2-diamine 95%
4-Methyl-5-nitrobenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₇H₉N₃O₂
MW: 167.17 g/mol
MDL Number: MFCD21604161
CAS Number: 65958-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR