H-Glu(OME)-OtBu·HCl 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-tert-Butyl 5-methyl 2-aminopentanedioate hydrochloride, H-Glu(OMe)-OtBu·HCl

A213635-1G A213635-250MG A213635-5G A213635-25G
76814-650EA 16 USD
76814-650 76814-652 77568-058 77671-904
H-Glu(OME)-OtBu·HCl 97%
H-Glu(OME)-OtBu·HCl

Precaución: For research use only.

Formula: C₁₀H₂₀ClNO₄
MDL Number: MFCD02094380
CAS Number: 34582-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR