(2-Oxo-2,3-dihydro-1H-indol-3-yl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Oxoindolin-3-yl)acetic acid

A213557-250MG A213557-1G A213557-100MG
76814-648EA 31.83 USD
76814-648 77612-100 77671-902
(2-Oxo-2,3-dihydro-1H-indol-3-yl)acetic acid 95%
(2-Oxo-2,3-dihydro-1H-indol-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
MDL Number: MFCD09035909
CAS Number: 2971-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR