Methyl 5-iodo-2-methylbenzoate 95%

Supplier: Ambeed
Warning

A213017-10G A213017-1G A213017-250MG A213017-5G A213017-100G A213017-25G
76814-638EA 40 USD
76814-638 76814-640 76814-642 76814-644 77883-519 77883-520
Methyl 5-iodo-2-methylbenzoate 95%
Methyl 5-iodo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉IO₂
MDL Number: MFCD11112847
CAS Number: 103440-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR