5,6-Dihydro-5-methyluracil 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methyldihydropyrimidine-2,4(1H,3H)-dione

A212622-100MG A212622-1G A212622-250MG
76814-626EA 72.15 USD
76814-626 76814-628 76814-630
5,6-Dihydro-5-methyluracil 97%
5,6-Dihydro-5-methyluracil

Precaución: for Research Use Only

Formula: C₅H₈N₂O₂
MW: 128.13 g/mol
MDL Number: MFCD00023159
CAS Number: 696-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR