2-Fluoro-4-hydroxyphenylacetic acid 95%

Supplier: Ambeed
Warning

A212604-5G A212604-1G A212604-250MG
76814-624EA 57 USD
76814-624 77612-088 77883-511
2-Fluoro-4-hydroxyphenylacetic acid 95%
2-Fluoro-4-hydroxyphenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MDL Number: MFCD09027748
CAS Number: 68886-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR