(1R,4R)-4-Ethyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane) 97%

Supplier: Ambeed
Warning

Synonyms: 4-Ethyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)

A212474-10G A212474-1G A212474-5G
76814-608EA 30.77 USD
76814-608 76814-610 76814-612
(1R,4R)-4-Ethyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane) 97%
(1R,4R)-4-Ethyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)

Precaución: for Research Use Only

Formula: C₂₀H₂₇F₃
MW: 324.42 g/mol
MDL Number: MFCD23115439
CAS Number: 2196180-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR