5-Thiazolecarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A212356-100G
A212356-10G
A212356-1G
A212356-250MG
A212356-25G
A212356-5G
76814-592EA
583.5
USD
76814-592
76814-594
76814-596
76814-598
76814-600
76814-602
5-Thiazolecarboxylic acid 97%
5-Thiazolecarboxylic acid
Precaución: for Research Use Only
Formula:
C₄H₃NO₂S MW: 129.1371 g/mol |
MDL Number:
MFCD03428539 CAS Number: 14527-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...