3-Nitro-5-(trifluoromethyl)pyridin-2-ol 98+%
Supplier: Ambeed
|
Warning
|
A212225-100G
A212225-1G
A212225-5G
76814-586EA
1289
USD
76814-586
76814-588
76814-590
3-Nitro-5-(trifluoromethyl)pyridin-2-ol 98+%
3-Nitro-5-(trifluoromethyl)pyridin-2-ol
Precaución: for Research Use Only
Formula:
C₆H₃F₃N₂O₃ MW: 208.09 g/mol |
MDL Number:
MFCD00153193 CAS Number: 1851650-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...