18-Bromo-6,9-octadecadiene 95%

Supplier: Ambeed
Warning

A211369-1G A211369-250MG
76814-550EA 138.98 USD
76814-550 76814-552
18-Bromo-6,9-octadecadiene 95%
18-Bromo-6,9-octadecadiene

Precaución: for Research Use Only

Formula: C₁₈H₃₃Br
MW: 329.35 g/mol
MDL Number: MFCD00034591
CAS Number: 4102-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR