Ethyl 2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate 97%

Supplier: Ambeed
Warning

A210520-1G A210520-5G
76814-524EA 108.22 USD
76814-524 76814-526
Ethyl 2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate 97%
Ethyl 2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD12090273
CAS Number: 261767-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR