N-Boc-4-oxo-L-proline-tert-butyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Bis(tert-butyl) (2S)-4-oxopyrrolidine-1,2-dicarboxylate, (S)-Di-tert-butyl 4-oxopyrrolidine-1,2-dicarboxylate, Boc-4-Oxo-Pro-OtBu

A210193-1G A210193-25G A210193-5G A210193-100G
76814-510EA 6.37 USD
76814-510 77568-026 77568-028 77593-370
N-Boc-4-oxo-L-proline-tert-butyl ester 97%
N-Boc-4-oxo-L-proline-tert-butyl ester

Precaución: For research use only.

Formula: C₁₄H₂₃NO₅
MW: 285.35 g/mol
MDL Number: MFCD06797013
CAS Number: 166410-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR