Dimethyl-3-oxoglutarate 95%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate

A208451-25G A208451-500G A208451-100G
76814-506EA 5 USD
76814-506 76814-508 77550-342
Dimethyl-3-oxoglutarate 95%
Dimethyl-3-oxoglutarate

Precaución: for Research Use Only

Formula: C₇H₁₀O₅
MDL Number: MFCD00008462
CAS Number: 1830-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR