5-Bromo-4-chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine 95%

Supplier: Ambeed
Danger

A207897-250MG A207897-1G
76814-496EA 20 USD
76814-496 77883-497
5-Bromo-4-chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine 95%
5-Bromo-4-chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₅BrClN₃
MDL Number: MFCD16249868
CAS Number: 1111237-76-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR