4-Chloro-2-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A207781-100G A207781-10G A207781-1G A207781-25G A207781-500G A207781-5G
76814-484EA 54 USD
76814-484 76814-486 76814-488 76814-490 76814-492 76814-494
4-Chloro-2-fluorobenzaldehyde 98%
4-Chloro-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO
MDL Number: MFCD00143282
CAS Number: 61072-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR