2-Iodo-6-methoxyaniline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Iodo-o-anisidine, 2-Amino-3-iodoanisole

A206606-100MG A206606-1G A206606-250MG A206606-5G
76814-462EA 18 USD
76814-462 76814-464 76814-466 76814-468
2-Iodo-6-methoxyaniline 98%
2-Iodo-6-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈INO
MDL Number: MFCD11977372
CAS Number: 354574-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR