Methyl 5-methyl-1,3,4-oxadiazole-2-carboxylate 95%

Supplier: Ambeed
Warning

A206259-100MG A206259-1G A206259-250MG
76814-456EA 16 USD
76814-456 76814-458 76814-460
Methyl 5-methyl-1,3,4-oxadiazole-2-carboxylate 95%
Methyl 5-methyl-1,3,4-oxadiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₅H₆N₂O₃
MDL Number: MFCD20921990
CAS Number: 37641-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR