10-Methoxy-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-one 98%
Supplier: Ambeed
|
Warning
|
A206157-25G
A206157-500G
76814-452EA
118
USD
76814-452
76814-454
10-Methoxy-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-one 98%
10-Methoxy-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-one
Precaución: for Research Use Only
Formula:
C₁₄H₁₀O₂S |
MDL Number:
MFCD00101207 CAS Number: 59743-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...