3-(Trifluoromethyl)azetidin-3-ol hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-Trifluoromethyl-3-azetidinol hydrochloride

A206115-100MG A206115-250MG A206115-5G A206115-1G
76814-446EA 13 USD
76814-446 76814-448 76814-450 77567-976
3-(Trifluoromethyl)azetidin-3-ol hydrochloride 98%
3-(Trifluoromethyl)azetidin-3-ol hydrochloride

Precaución: For research use only.

Formula: C₄H₇ClF₃NO
MDL Number: MFCD13248564
CAS Number: 848192-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR