3-Bromo-5-methylpicolinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-methylpyridine-2-carboxylic acid

A204780-10G A204780-1G A204780-250MG A204780-25G A204780-5G
76814-414EA 59 USD
76814-414 76814-416 76814-418 76814-420 76814-422
3-Bromo-5-methylpicolinic acid 97%
3-Bromo-5-methylpicolinic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MDL Number: MFCD17214206
CAS Number: 1211515-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR