N-[5-Bromomethyl-4-(4-fluorophenyl)-6-isopropylpyrimidine-2-yl]-N-methylmethane sulfonamide 97%

Supplier: Ambeed
Danger

A204526-100G A204526-10G A204526-1G A204526-250MG A204526-25G A204526-5G
76814-394EA 105.03 USD
76814-394 76814-396 76814-398 76814-400 76814-402 76814-404
N-[5-Bromomethyl-4-(4-fluorophenyl)-6-isopropylpyrimidine-2-yl]-N-methylmethane sulfonamide 97%
N-[5-Bromomethyl-4-(4-fluorophenyl)-6-isopropylpyrimidine-2-yl]-N-methylmethane sulfonamide

Precaución: for Research Use Only

Formula: C₁₆H₁₉BrFN₃O₂S
MW: 416.30 g/mol
MDL Number: MFCD08694308
CAS Number: 799842-07-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR