H-β-(3-Benzothienyl)-Ala-OH 97%

Supplier: Ambeed
Warning

Synonyms: H-3-BenzoThienyl-Ala-OH, (S)-2-Amino-3-(benzo[b]thiophen-3-yl)propanoic acid, 3-(Benzo[b]thiophen-3-yl)-L-alanine

A203774-10G A203774-1G A203774-250MG A203774-5G
76814-378EA 399 USD
76814-378 76814-380 76814-382 76814-384
H-β-(3-Benzothienyl)-Ala-OH 97%
H-β-(3-Benzothienyl)-Ala-OH

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂S
MDL Number: MFCD00079684
CAS Number: 72120-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR