4-Iodo-N-methylaniline hydrochloride 95%

Supplier: Ambeed
Warning

A203662-1G A203662-250MG A203662-5G
76814-368EA 60.48 USD
76814-368 76814-370 76814-372
4-Iodo-N-methylaniline hydrochloride 95%
4-Iodo-N-methylaniline hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉ClIN
MW: 269.51 g/mol
MDL Number: MFCD12912932
CAS Number: 84250-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR