1-Bromo-4-(phenylsulfonyl)benzene 95%

Supplier: Ambeed
Warning

A203189-100MG A203189-10G A203189-1G A203189-250MG A203189-5G A203189-25G
76814-352EA 5 USD
76814-352 76814-354 76814-356 76814-358 76814-360 77883-471
1-Bromo-4-(phenylsulfonyl)benzene 95%
1-Bromo-4-(phenylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₁₂H₉BrO₂S
MDL Number: MFCD00034047
CAS Number: 23038-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR