1-Bromo-4-(isopropylsulfonyl)benzene 95%

Supplier: Ambeed
Warning

A202981-100MG A202981-1G A202981-250MG
76814-346EA 10.61 USD
76814-346 76814-348 76814-350
1-Bromo-4-(isopropylsulfonyl)benzene 95%
1-Bromo-4-(isopropylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂S
MW: 263.15 g/mol
MDL Number: MFCD20441891
CAS Number: 70399-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR