6-Chloro-5-fluoro-2-methylpyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A202973-100MG A202973-1G A202973-250MG A202973-5G
76814-338EA 9.55 USD
76814-338 76814-340 76814-342 76814-344
6-Chloro-5-fluoro-2-methylpyrimidin-4-amine 95%
6-Chloro-5-fluoro-2-methylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₅ClFN₃
MW: 161.565 g/mol
MDL Number: MFCD11846146
CAS Number: 943006-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR