N-(2-Hydroxy-3-(piperidin-1-yl)propoxy)nicotinimidamide dihydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: BGP-15

A202611-10MG A202611-1MG A202611-25MG A202611-50MG A202611-5MG
76814-324EA 185.66 USD
76814-324 76814-326 76814-328 76814-330 76814-332
N-(2-Hydroxy-3-(piperidin-1-yl)propoxy)nicotinimidamide dihydrochloride 95%
N-(2-Hydroxy-3-(piperidin-1-yl)propoxy)nicotinimidamide dihydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₄Cl₂N₄O₂
MW: 351.27 g/mol
MDL Number: MFCD21603901
CAS Number: 66611-37-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR