6-Bromochroman-2-one 97%

Supplier: Ambeed
Warning

A201571-1G A201571-250MG A201571-5G
76814-314EA 82.76 USD
76814-314 76814-316 76814-318
6-Bromochroman-2-one 97%
6-Bromochroman-2-one

Precaución: for Research Use Only

Formula: C₉H₇BrO₂
MW: 227.05 g/mol
MDL Number: MFCD13659375
CAS Number: 20921-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR