4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide 98+%

Supplier: Ambeed
Warning

A201295-10MG A201295-1MG A201295-50MG A201295-5MG
76814-300EA 63 USD
76814-300 76814-302 76814-304 76814-306
4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide 98+%
4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide

Precaución: for Research Use Only

Formula: C₁₆H₂₅N₃O₃
MDL Number: MFCD03453554
CAS Number: 251456-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR