trans-4-Propyl-4'-(2,3,4-trifluorophenyl)-1,1'-bi(cyclohexane) 97%

Supplier: Ambeed
Warning

A201258-1G A201258-5G
76814-296EA 10.61 USD
76814-296 76814-298
trans-4-Propyl-4'-(2,3,4-trifluorophenyl)-1,1'-bi(cyclohexane) 97%
trans-4-Propyl-4'-(2,3,4-trifluorophenyl)-1,1'-bi(cyclohexane)

Precaución: for Research Use Only

Formula: C₂₁H₂₉F₃
MW: 338.45 g/mol
MDL Number: MFCD23115464
CAS Number: 139056-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR