tert-Butyl (3-(methylamino)phenyl)carbamate 98%

Supplier: Ambeed
Warning

A200767-100MG A200767-250MG A200767-5G A200767-1G
76814-282EA 47 USD
76814-282 76814-284 76814-286 77883-460
tert-Butyl (3-(methylamino)phenyl)carbamate 98%
tert-Butyl (3-(methylamino)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MDL Number: MFCD11858352
CAS Number: 1134328-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR