Methyl 3-fluoro-4-formylbenzoate 98%

Supplier: Ambeed
Warning

A200481-1G A200481-5G A200481-25G A200481-250MG
76814-272EA 40.32 USD
76814-272 76814-274 77567-896 77671-786
Methyl 3-fluoro-4-formylbenzoate 98%
Methyl 3-fluoro-4-formylbenzoate

Precaución: For research use only.

Formula: C₉H₇FO₃
MW: 182.148 g/mol
MDL Number: MFCD16036860
CAS Number: 74733-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR