L-Homocysteine 95%

Supplier: Ambeed
Danger

Synonyms: (S)-2-Amino-4-mercaptobutanoic acid, H-HoCys-OH

A200253-1G A200253-1MG A200253-25MG A200253-250MG A200253-100MG A200253-5G
76814-254EA 309.79 USD
76814-254 76814-256 76814-258 77567-886 77611-936 77611-938
L-Homocysteine 95%
L-Homocysteine

Precaución: For research use only.

Formula: C₄H₉NO₂S
MW: 135.19 g/mol
MDL Number: MFCD00151320
CAS Number: 6027-13-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR