1,5-Dichloro-3-methyl-2-nitrobenzene 95%

Supplier: Ambeed
Warning

A200026-10G A200026-250MG A200026-5G
76814-236EA 104 USD
76814-236 76814-238 76814-240
1,5-Dichloro-3-methyl-2-nitrobenzene 95%
1,5-Dichloro-3-methyl-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD11878022
CAS Number: 118665-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR