N-Benzyl-2-chloro-9H-purin-6-amine 95%

Supplier: Ambeed
Warning

A199508-1G A199508-250MG
76814-228EA 119.89 USD
76814-228 76814-230
N-Benzyl-2-chloro-9H-purin-6-amine 95%
N-Benzyl-2-chloro-9H-purin-6-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClN₅
MW: 259.69 g/mol
MDL Number: MFCD09701466
CAS Number: 39639-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR