(1S,4R)-4-Aminocyclopent-2-enecarboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A199298-100MG A199298-1G A199298-250MG
76814-204EA 70.02 USD
76814-204 76814-206 76814-208
(1S,4R)-4-Aminocyclopent-2-enecarboxylic acid hydrochloride 95%
(1S,4R)-4-Aminocyclopent-2-enecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀ClNO₂
MW: 163.60 g/mol
MDL Number: MFCD11504996
CAS Number: 130931-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR