(E)-3-(4-Amino-3,5-dimethylphenyl)acrylonitrile hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(4-Amino-3,5-dimethylphenyl)-2-propenenitrile hydrochloride

A198712-1G A198712-250MG A198712-5G
76814-182EA 68.96 USD
76814-182 76814-184 77671-758
(E)-3-(4-Amino-3,5-dimethylphenyl)acrylonitrile hydrochloride 95%
(E)-3-(4-Amino-3,5-dimethylphenyl)acrylonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂·HCl
MW: 208.69 g/mol
MDL Number: MFCD18711816
CAS Number: 661489-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR