5-Methoxybenzo[d]thiazole-2-carbonitrile 95%

Supplier: Ambeed
Warning

A198575-100MG A198575-1G A198575-250MG
76814-172EA 95 USD
76814-172 76814-174 76814-176
5-Methoxybenzo[d]thiazole-2-carbonitrile 95%
5-Methoxybenzo[d]thiazole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂OS
MDL Number: MFCD11217490
CAS Number: 7267-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR