2-Cyano-4-fluorophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2-Cyano-4-fluorophenyl)boronic acid

A197879-100MG A197879-5G A197879-1G A197879-250MG
76814-154EA 13 USD
76814-154 77883-439 77883-437 77883-438
2-Cyano-4-fluorophenylboronic acid 95%
2-Cyano-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BFNO₂
MDL Number: MFCD12546526
CAS Number: 876601-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR