3-Methyl-1H-pyrazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methylpyrazole-5-carboxylic acid

A197553-100G A197553-1G A197553-25G A197553-5G
76814-136EA 583.5 USD
76814-136 76814-138 76814-140 76814-142
3-Methyl-1H-pyrazole-5-carboxylic acid 95%
3-Methyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
MDL Number: MFCD00462235
CAS Number: 696-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR