2-Phenyl-5-(trifluoromethyl)oxazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A197165-1G A197165-5G
76814-126EA 77.45 USD
76814-126 76814-128
2-Phenyl-5-(trifluoromethyl)oxazole-4-carboxylic acid 95%
2-Phenyl-5-(trifluoromethyl)oxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₆F₃NO₃
MW: 257.17 g/mol
MDL Number: MFCD00203251
CAS Number: 236736-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR