4-Amino-2-chloro-3-fluoropyridine 96%

Supplier: Ambeed
Warning

A196037-100MG A196037-10G A196037-1G A196037-250MG A196037-25G A196037-5G A196037-100G
76814-098EA 5 USD
76814-098 76814-100 76814-102 76814-104 76814-106 76814-108 77883-422
4-Amino-2-chloro-3-fluoropyridine 96%
4-Amino-2-chloro-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄ClFN₂
MDL Number: MFCD16606117
CAS Number: 1227577-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR