trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl (1R,4R)-4-(hydroxymethyl)cyclohexane-1-carboxylate, Methyl trans-4-hydroxymethylcyclohexanecarboxylate, (1R,4R)-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate

A195875-100MG A195875-1G A195875-250MG A195875-25G A195875-5G
76814-076EA 19.1 USD
76814-076 76814-078 76814-080 76814-082 76814-084
trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate 97%
trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₆O₃
MW: 172.22 g/mol
MDL Number: MFCD03840884
CAS Number: 110928-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR