Pyrimethamine 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 5-(4-Chlorophenyl)-6-ethylpyrimidine-2,4-diamine, 5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine

A195764-100MG A195764-10G A195764-1G A195764-25G A195764-5G A195764-100G
76814-060EA 5.31 USD
76814-060 76814-062 76814-064 77611-884 77611-886 77671-738
Pyrimethamine 98% (by HPLC)
Pyrimethamine

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClN₄
MW: 248.71 g/mol
MDL Number: MFCD00057350
CAS Number: 58-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR